Alfa Aesar™ Stainless Steel powder, -10+20 mesh, Type 316-L

Manufacturer: Alfa Aesar™ 43458A3

Quantity 2kg

 View more versions of this product

Catalog No. AA43458A3

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel powder, Type 316-L

    Specifications

    Quantity 2kg
    Health Hazard 2 GHS H Statement
    H372-H351-H319-H317-H335
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    Health Hazard 1 Danger
    Mesh Size −10+20 Mesh
    Color Gray
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
    Density 1.57g/mL
    Chemical Name or Material Stainless Steel powder, Type 316-L

    GHS H Statement
    H372-H351-H319-H317-H335
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit