Alfa Aesar™ Stainless Steel powder, -10+25 mesh, Type 316-L

Manufacturer: Alfa Aesar™ 4345936

Quantity 500g

 View more versions of this product

Catalog No. AA4345936

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel powder, Type 316-L

    Specifications

    Quantity 500g
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00148499
    Mesh Size −10+25 Mesh
    Chemical Name or Material Stainless Steel powder, Type 316-L

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit