Learn More
Alfa Aesar™ Stainless Steel powder, -100 mesh, annealed, Type 410-L
Manufacturer:  Alfa Aesar™ 11088A3
Quantity 2kg
 View more versions of this product
Catalog No. AA11088A3
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel powder, Type 410-L, Annealed
Specifications
Fe:Cr; 87.5:12.5 wt%
2kg
MFCD00148499
−100 Mesh
Powder
Stainless Steel powder, Type 410-L, Annealed
Safety and Handling
SDS & Safety Documents
TSCA : TSCA