Learn More
Alfa Aesar™ Stainless Steel powder, -100 mesh, Type 304-L
Manufacturer:  Alfa Aesar™ 11085A3
Quantity 2kg
 View more versions of this product
Catalog No. AA11085A3
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel powder, Type 304-L
Specifications
Fe:Cr:Ni; 70:19:11 wt%
2kg
MFCD00148499
−100 Mesh
Powder
Stainless Steel powder, Type 304-L
Safety and Handling
SDS & Safety Documents
TSCA : TSCA