Learn More
Stainless Steel powder, -100 mesh, Type 304-L, Thermo Scientific Chemicals
$21.55 - $249.65
Specifications
Chemical Name or Material | Stainless Steel powder, Type 304-L |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Mesh Size | −100 Mesh |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA1108518
|
Thermo Scientific Chemicals
01108518 |
50 g |
Each for $21.55
Order in Bulk
Request bulk or custom formats |
|
|||||
AA1108536
|
Thermo Scientific Chemicals
01108536 |
500 g |
Each
for $84.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA11085A3
|
Thermo Scientific Chemicals
011085A3 |
2 kg |
Each
for $249.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Stainless Steel powder, -100 mesh, Type 304-L is the most common stainless steel alloy widely used in wire weaving. It has excellent corrosion resistance and can withstand temperatures of 1100 degrees F. It has better welding characteristics for lower carbon content.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 304-L | |
MFCD00148499 | |
−100 Mesh |
Fe:Cr:Ni; 70:19:11 wt% | |
Insoluble in water. | |
Powder |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.