Learn More
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01108936
Description
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
500 g | |
−100 Mesh |
Stainless Steel powder, Type 316-L | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
Safety and Handling
GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only