Learn More
Alfa Aesar™ Stainless Steel powder, -140 mesh, Type 303-L
Manufacturer:  Alfa Aesar™ 1108430
 View more versions of this product
Catalog No. AA1108430
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
Specifications
Specifications
Fe:Cr:Ni; 70:17:13 wt%
250 g
−140 Mesh
Powder
Stainless Steel powder, Type 303-L
MFCD00148499
Insoluble in water.
Safety and Handling
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures