Alfa Aesar™ Stainless Steel powder, -140 mesh, Type 303-L

Manufacturer: Alfa Aesar™ 1108418

Quantity 50g

 View more versions of this product

Catalog No. AA1108418

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel powder, Type 303-L

    Specifications

    Quantity 50g
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:17:13 wt%
    Mesh Size −140 Mesh
    Chemical Name or Material Stainless Steel powder, Type 303-L

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit