Learn More
Alfa Aesar™ Stainless Steel powder, -16+25 mesh, Type 316-L
Manufacturer:  Alfa Aesar™ 4345736
Quantity 500g
 View more versions of this product
Catalog No. AA4345736
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel powder, Type 316-L
Specifications
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
500g
MFCD00148499
−16+25 Mesh
Powder
Stainless Steel powder, Type 316-L
Safety and Handling
SDS & Safety Documents
GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : TSCA