Learn More
Alfa Aesar™ Stainless Steel powder, -20+35 mesh, Type 316-L
Manufacturer:  Alfa Aesar™ 4346036
Quantity 500g
 View more versions of this product
Catalog No. AA4346036
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel powder, Type 316-L
Specifications
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
500g
MFCD00148499
−20+35 Mesh
Powder
Stainless Steel powder, Type 316-L
Safety and Handling
SDS & Safety Documents
TSCA : TSCA