Learn More
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
$67.70 - $410.65
Specifications
Chemical Name or Material | Stainless Steel powder, Type 316-L |
---|---|
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA8839022
|
Thermo Scientific Chemicals
08839022 |
100 g |
Each for $67.70
Order in Bulk
Request bulk or custom formats |
|
|||||
AA8839036
|
Thermo Scientific Chemicals
08839036 |
500 g |
Each
for $141.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA88390A3
|
Thermo Scientific Chemicals
088390A3 |
2 kg |
Each
for $410.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 316-L | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Insoluble in water. | |
Powder |
Gray | |
MFCD00148499 | |
−325 Mesh |
Safety and Handling
GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.