Alfa Aesar™ Stainless Steel powder, -325 mesh, Type 430-L

Manufacturer: Alfa Aesar™ 47290A3

Quantity 2kg

 View more versions of this product

Catalog No. AA47290A3

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel, Type 430-L

    Specifications

    Quantity 2kg
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00148499
    Health Hazard 2 GHS H Statement
    H351-H319-H335
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Mesh Size −325 Mesh
    TSCA TSCA
    Physical Form Powder
    Health Hazard 1 Warning
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%
    Chemical Name or Material Stainless Steel, Type 430-L

    GHS H Statement
    H351-H319-H335
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit