Alfa Aesar™ Stainless Steel powder, -325 mesh, Type 430-L

Manufacturer: Alfa Aesar™ 4729022

Quantity 100g

 View more versions of this product

Catalog No. AA4729022

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel, Type 430-L

    Specifications

    Quantity 100g
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00148499
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H319-H335
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%
    Mesh Size −325 Mesh
    Chemical Name or Material Stainless Steel, Type 430-L

    GHS H Statement
    H351-H319-H335
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit