Learn More
Alfa Aesar™ Stainless Steel powder, -40+80 mesh, Type 316
Manufacturer:  Alfa Aesar™ 42944A1
Quantity 1kg
 View more versions of this product
Catalog No. AA42944A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel powder, Type 316
Specifications
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
1kg
MFCD00148499
−40+80 Mesh
Powder
Stainless Steel powder, Type 316
Safety and Handling
SDS & Safety Documents
TSCA : TSCA