Stainless-Steel powder, APS 15 micron, Type 430-L, Alfa Aesar™

Manufacturer: Alfa Aesar 45516A3

Quantity 2kg

 View more versions of this product

Catalog No. AA45516A3

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Physical Form Powder
    Molecular Formula Fe: 82.9; Cr: 16.5; Si: 0.5; C: 0.02 wt%
    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel, Type 430-L

    Specifications

    Quantity 2kg
    Color Silver-gray
    Physical Form Powder
    Odor Odorless
    Molecular Formula Fe: 82.9; Cr: 16.5; Si: 0.5; C: 0.02 wt%
    MDL Number MFCD00148499
    Appearance Silver gray powder
    Health Hazard 1 H351: Suspected of causing cancer.
    Chemical Name or Material Stainless Steel, Type 430-L

    H351: Suspected of causing cancer.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit