Alfa Aesar™ Stainless Steel rod, 9.5mm (0.375 in.) dia., Type 303

Manufacturer: Alfa Aesar™ 42707BV

Quantity 30cm

 View more versions of this product

Catalog No. AA42707BV

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel rod, Type 303, 9.5mm (0.375 in.) dia.

    Specifications

    Quantity 30cm
    TSCA TSCA
    Physical Form Rod
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 73:18:9 wt%
    Chemical Name or Material Stainless Steel rod, Type 303, 9.5mm (0.375 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit