Stainless Steel Spheres, 1mm (0.04 in.) dia., Alfa Aesar™

Manufacturer: Alfa Aesar 42018KN

Quantity 100 Pieces

 View more versions of this product

Catalog No. AA42018KN

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Spheres, 1mm (0.04 in.) dia.
    Molecular Formula Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel

    Specifications

    Quantity 100 Pieces
    Physical Form Spheres, 1mm (0.04 in.) dia.
    Molecular Formula Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
    MDL Number MFCD00148499
    Appearance Spheres
    Health Hazard 1 H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
    Chemical Name or Material Stainless Steel

    H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit