Learn More
Stainless Steel spheres, 1mm (0.04in) dia, Type 316, Thermo Scientific Chemicals
$34.65 - $69.65
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Appearance | Spheres |
Molecular Formula | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Form | Sphere |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA42018KM
|
Thermo Scientific Chemicals
042018KM |
25 Pc. |
Pack of 25
for $34.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA42018KN
|
Thermo Scientific Chemicals
042018KN |
100 Pc. |
Pack of 100
for $69.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Sphere |
Spheres | |
MFCD00148499 |
Safety and Handling
H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.