Alfa Aesar™ Stainless Steel spheres, 2mm (0.08 in.) dia., Type 302

Manufacturer: Alfa Aesar™ 42016KN

Quantity 100 pieces

 View more versions of this product

Catalog No. AA42016KN

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel spheres, Type 302, 2mm (0.08 in.) dia.

    Specifications

    Quantity 100 pieces
    TSCA TSCA
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mn:Si; 70:18:9:2:1 wt%
    Chemical Name or Material Stainless Steel spheres, Type 302, 2mm (0.08 in.) dia.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit