Learn More
Stainless Steel spheres, 3mm (0.12in) dia, Type 316, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Stainless Steel spheres, Type 316 |
---|---|
Name Note | 3mm (0.12 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA42020KM
|
Thermo Scientific Chemicals
042020KM |
25 Pc. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
AA42020KN
|
Thermo Scientific Chemicals
042020KN |
100 Pc. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel spheres, Type 316 | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Insoluble in water. |
3mm (0.12 in.) dia. | |
MFCD00148499 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.