Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo™ Stand Rod 28 in. (711 mm) x 1 in. (25 mm)

28 in. (711 mm) length Support Rod compatible with Caframo A110BASE (14-638-265) Heavy-Duty stand base.

Manufacturer:  Caframo™ ROD28

Catalog No. 14-501-092


Add to cart

Description

Description

  • Replacement rod for standard A110 Stand (14-500-354 and 14-500-2)
  • Also compatible with Caframo A110BASE (14-638-265) Heavy-Duty stand base
  • Composed of 304 stainless steel tub, 1 in (25 mm) dia
Specifications

Specifications

1 in.
Stand rod only
711 mm
25 mm
28 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit