Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Stainless Steel Syringe Needles

Long needles feature a female luer lock connector and beveled non-coring tip, designed for use with solvent retrieval.

Manufacturer:  Synthware™ S389300

 View more versions of this product

Catalog No. 31-502-330


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Long, stainless steel syringe needles
  • Female luer lock connector
  • Beveled non-coring tip
  • Compatible with both luer slip, and luer lock syringes
  • For use with solvent retrieval
Specifications

Specifications

0.90 mm
Yes
300 mm
Luer Lock/Luer Slip Syringes
No
Stainless Steel
Solvent Retrieval
Syringe Needle
Beveled Tip
Luer Lock
No
Non-sterile
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit