Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Stainless Steel Tall Cabinet Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Stainless Steel furnishings are designed specifically for research and healthcare laboratories.
 View more versions of this product
Catalog No. 06-000-577
Description
Description
  • Stainless Steel Tall Cabinets come standard with locks and 5 adjustable shelves.
  • Must be anchored to the wall
  • Made of Type #304 stainless steel with #4 finish.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Stainless Steel
55.9 cm
84.6 in.
Left and Right
Stainless Steel
5
48 in
22 in.
Solid Door
214.9 cm
Locks
2
2 years
121.9 cm