Alfa Aesar™ Stainless Steel tubing, 0.82mm (0.032in) OD, 0.51mm (0.02in) ID, Type 304

Manufacturer: Alfa Aesar™ 13468CE

Quantity 5 x 30cm

 View more versions of this product

Catalog No. AA13468CE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel tubing, Type 304, OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)

    Specifications

    Quantity 5 x 30cm
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:19:11 wt%
    TSCA TSCA
    Chemical Name or Material Stainless Steel tubing, Type 304, OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit