Learn More
Thermo Scientific™ Stainless Steel Unions, Tees and Crosses Available on GSA/VA Contract for Federal Government customers only.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Manufacturer:  Thermo Scientific™ U438
 View more versions of this product
Catalog No. 03-170-446
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific? Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
Description & Specifications
Specifications
HPLC
LC
~0.0?L
Union
Upchurch, Parker; ~0.0uL Swept Volume