Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Stainless Steel wire, 0.127mm (0.005 in.) dia., Type 304

Filtration, electrodes

Manufacturer:  Alfa Aesar™ 40944BZ

 View more versions of this product

Catalog No. AA40944BZ


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

Specifications

Specifications

Fe:Cr:Ni; 70:19:11 wt%
150 cm
MFCD00148499
Odorless
Stainless Steel wire, Type 304
0.127mm (0.005 in.) dia.
Insoluble in water.
Wire
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit