Learn More
Thermo Scientific™ StainMate™ Max 10 Batch Stainer Accessories
Maintain Thermo Scientific™ StainMate™ Max 10 Batch Stainer with StainMate Max 10 Batch Stainer Accessories.
$602.00
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
E103SLEXTB
View Documents
Thermo Scientific™
E103/SL/EXTB
External dish/bath Each for $602.00
Add to cart
Description
Description
Maintain Thermo Scientific™ StainMate™ Max 10 Batch Stainer with StainMate Max 10 Batch Stainer Accessories.
Accessories for StainMate Max 10 Batch Stainer include:
  • Lid
  • Fume hood
  • Pinth with drawer
  • Bath dish for Belair model
  • Additional dish/bath
  • Water dish/bath
  • External dish/bath
  • Drying module
  • Rack adapter
  • Additional Rack for up to 20 slides
  • Additional Rack for up to 30 slides
Dryer Module
  • Fan-assisted slide dryer operates at ∽70°C
  • Hooks onto either the left- or right-hand side of the StainMate module
Fume Hood
  • Extracts fumes through a replaceable charcoal filter
  • Fits easily inside the base of the hood
  • Transparent door is hinged for easy access
External Dish/Bath
  • Attach to either side of the StainMate module to increase the capacity
  • Constructed of polished stainless steel with white acetal bath and lid
  • Capacity: 38mL