Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stains-All, 95%

Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids

$51.10 - $188.50

Specifications

Assay 0.95
CAS 7423-31-6
Formula Weight 559.58g/mol
Storage Requirements 2° to 8°C
Format Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6263006 View Documents Alfa Aesar™
J6263006
5g Each for $188.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6263003 View Documents Alfa Aesar™
J6263003
1g Each for $51.10
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

0.95
559.58g/mol
Powder
Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids
Hematology Stain
7423-31-6
2° to 8°C
Light sensitive
Dark Gray or Green
Safety and Handling

Safety and Handling

EINECSNumber : 231-047-7

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit