Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAM 1, 2 Sheep anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
$153.04 - $436.00
Specifications
Antigen | STAM 1, 2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Description
STAM 1, 2 Polyclonal specifically detects STAM 1, 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
STAM 1, 2 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Sterile water with No Preservative | |
8027 | |
A synthetic peptide from the internal part of human STAM 1, 2 (STAM-1, STAM-2) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in many species including rat, mouse, zebra fish and xenopus. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. | |
Appears to be specific for STAM1 & 2. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Sheep | |
Human, Mouse, Rat | |
DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 | |
STAM | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title