Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ LER Linearity Check (6x4mL)
Designed for maximum stability and ease-of-use
Supplier: Stanbio™ 2450604
This item is not returnable.
View return policy
Description
- Enzymatic beta-hydroxybutyrate reaction
- Ready-to-use liquid reagents
- Use serum or plasma
- Linear up to 4.5 mmol/L
- Read at 505 nm
Specifications
CE Marked | |
Quantitative testing | |
Liquid |
LER Linearity Check | |
6 x 4 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction