Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Calibrator Set For Glycated Serum Protein Assay

For Glycated Serum Protein LiquiColor™ Assay

Manufacturer:  Stanbio™ 2360401

Catalog No. SB2360401


Add to cart
This item is not returnable. View return policy

Description

Description

Contents: Calibrator 1 - 2 vials, Calibrator 2 - 2 vials, 4 x 1mL

Specifications

Specifications

Powder
Cal 1: 2 x 1 mL; Cal 2: 2 x 1 mL
Yes
Glycated Serum Protein (GSP) calibrator
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit