Learn More
Stanbio™ STAT-Site™ M BHB Diluent (6 x 3mL Vials)
GSA_VA
Assurance from false-positives, producing more confidence in results and greater test utility
$175.70 - $456.50
Specifications
For Use With (Application) Blood Ketone testing
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Type Includes Price Quantity & Availability  
SB202000
View Documents
Stanbio™
202000
6 x 3 mL STAT-Site M BHB Diluent STAT-Site M BHB Diluent
Each for $456.50
Add to cart
SB303500
GSA_VA
View Documents
Stanbio™
303500
1 x 1 mL STAT-Site M BHB Linearity STAT-Site M BHB Linearity
Each for $72.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
SB303000
GSA_VA
View Documents
Stanbio™
303000
2 x 3 mL STAT-Site M BHB Controls STAT-Site M BHB Controls
Each for $175.70
Add to cart
Description
Description
  • Quantitative testing–results in two minutes
  • Metabolic status, insulin uptake, and end of catabolism
  • STAT differential diagnosis between lacticacidosis and ketoacidosis
  • Blood ketone testing for diagnosis and monitoring of ketoacidosis
  • Tests for Β-hydroxybutyrate, the most accurate marker for measuring Diabetic Ketoacidosis (DBA)
  • Monitors the status of ketoacidosis, documents insulin uptake and defines the end of catabolism
  • An alternative to qualitative nitroprusside tests
  • Virtually no interference from captopril and other sulfhydryl drugs
  • Can also aid in STAT differential diagnosis between lacticacidosis and ketoacidosis
  • Use with Stanbio Stat-Site™ Analyzer for convenient, bedside testing
Specifications
Specifications
Blood Ketone testing