Learn More
Stanbio™ STAT-Site™ Mβ-HB Meter
Measure β-hydroxybutyrate quickly and quantitatively
Specifications
Quantity Each
View More Specs
Products 2
Catalog Number Mfr. No. Precision Type Price Quantity & Availability  
Catalog Number Mfr. No. Precision Type Price Quantity & Availability  
SB400400
View Documents
Stanbio™
400400
Low control: 6.1% CV/High control: 3.9% CV Ketone Meter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
SB400400SP
View Documents
Stanbio™
400400SP
NA Ketone Testing Starter Kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Determine the presence and degree of ketosis using only 10μL of serum or plasma.
  • β-hydroxybutyrate accounts for 78% of the ketone bodies
  • Meter provides results in less than 80 seconds with a 10μL sample
  • Rapid testing reduces time and costs in intensive care setting
  • Precise enough to follow the progression of treatment
  • Simply place test strip in meter, apply sample and read result
  • Saves space on lab bench
Order Info
Test Strips come in packs of 40: four canisters of 10 strips (see Cat. No. SB401010).
Specifications
Specifications
Each