Learn More
Stanbio™ RELY™ H. Pylori Rapid Test Kit
For diagnostic use
Manufacturer:  Stanbio™ 6300020
Catalog No. 22-022-517
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Two-step results in 10 minutes
  • CLIA waived for whole blood samples
  • Suitable for use with whole blood, serum, or plasma
Specifications
Specifications
Room Temperature
Identification of H. pylori
H. pylori Rapid Test Kit
20 Cassettes
Waived
Cassette
Procedural and external control