Learn More
Stanbio™ True™ 20 Plus hCG Pregnancy Test Set
For diagnostic use
Manufacturer:  Stanbio™ 144050
Catalog No. 22-022-520
Add to cart
Description
Description
  • Provides fast results
  • Detects 20mIU/mL hCG for both urine and serum samples
Certifications
CLIA waived
Specifications
Specifications
20mIU/mL
10 sec.
Serum, Urine
>99%
hCG pregnancy test kit
>99%
Waived
Room Temperature
hCG in Serum or Urine
50 Tests