Learn More
Cole-Parmer Meiji Techno PBH STAND Standard Microscope Stand; Illumination Source, 6 V, 10 W Halogen Bulb; 115V  DFS Item
Manufacturer:  Cole-Parmer 4840420
Features both incident (top) and transmitted (bottom) illumination. A transformer with selector switch is built into the base, allowing the lights to be used separately or simultaneously.
Catalog No. 50-631-688
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.