Learn More
Research Products International CorpSupplier Diversity Partner Disposable Plastic Cuvette, Spectrophotometer Standard, 10 mm Pathlength, 3.5 ml, 1000 per Case DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 248964
High quality rectangular cuvettes for UV/VIS spectrophotometers. Spectrophotometer cuvettes feature two polished windows and available with standard interior capacity or a semi-micro size for reduced sample volumes. Cuvettes offered in the most common pathlength-recommeded wavelength is 340-800 nm. Packaged 100 per tray, ten trays (1000 cuvettes) per case. Caps available separately.
Catalog No. 50-476-565