Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
Manufacturer: Thermo Scientific™ 365CL221
 View more versions of this product
Catalog No. 36-5CL-221
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Injector
    Type HPLC Autosampler Syringe
    Disposable No
    Item Description 5uL
    Volume (Metric) 5µL
    Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
    • Square tip to prevent damage to the injector
    • Wide range of volumes
    • Precision made from borosilicate glass and stainless steel
    • Robust design and easy-to-read markings
  • Description & Specifications
Related Products
Product Content Feedback