Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Adaptor Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Supplier: Thermo Scientific™ SH31088.01
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Approved for storage at ambient conditions (15°C to 30°C). | |
6.35mm (1/4 in) x 1.6mm (1/16 in) wall to 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.