Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 601
Used for clarifying liquids and rapid filtration of coarse precipitates: lead sulfate, lead chromate, calcium oxalate/carbonate.
Supplier:  Ahlstrom-Munksjo 60100900
 View more versions of this product
Catalog No. 09-790-55D
Explore available promotions
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Disc
Medium
601
2.5 μm
Clarifying liquids, rapid filtration of coarse precipitates: lead sulfate, lead chromate, calcium oxalate/carbonate, aluminum phosphate
9 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.