Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 631
A medium speed grade used for the filtration of coarse particles and gelatinous precipitates such as ferric aluminum and magnesium hydroxides. It is also used in air monitoring and gas filtration when high flow rate is needed.
Manufacturer: Ahlstrom-Munksjo 63102400
Specifications
10μm | |
Filtration of coarse particles and gelatinous precipitates; air monitoring and gas filtration when high flow rate is needed | |
24 cm |
Medium | |
631 |