Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 642

A medium speed grade used in general laboratory applications to retain particles above 2 μm in size. It is also used in agricultural and environmental laboratories for plant growth trials, soil testing and air analysis for monitoring specific contaminants.

$59.90 - $239.00

Specifications

Particle Retention 2 μm
Flow Rate Slow
For Use With (Application) General lab applications; agricultural and environmental labs for plant growth trials, soil testing, and air analysis for monitoring contaminants
Grade 642
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09-790-70D
View Documents
Ahlstrom-Munksjo
64201100
11 cm
Pack of 100 for $59.90
Add to Cart
 
22-293721
View Documents
Ahlstrom-Munksjo
64202500
25 cm
Pack of 100 for $239.00
Add to Cart
 
Description

Description

Specifications

Specifications

2 μm
Slow
General lab applications; agricultural and environmental labs for plant growth trials, soil testing, and air analysis for monitoring contaminants
642
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.