Standard Reference Solution, pH 4.01 Buffer, Color Coded Red, Macron Fine Chemicals™

macron

Manufacturer: Macron 009710

Quantity 4L

 View more versions of this product

Catalog No. 14-650-402

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid

    Specifications

    Quantity 4L
    Physical Form Liquid
    Appearance Clear, red solution
    pH Range 4.00 to 4.02
    Traceability to NIST Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit