Standard Reference Solution, pH 7.0 Buffer, Color Coded Yellow, Macron Fine Chemicals™


Manufacturer: AVANTOR 009810/DEL

 View all options for this product

Catalog No. 14-650-403


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

    Chemical Identifiers

    Physical Form Liquid


    Appearance Clear, yellow solution
    pH Range 6.96 to 7.04
    Traceability to NIST Yes
    Physical Form Liquid

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit