Learn More
Description
Used when the filtered serum can be immediately poured off for analysis.
Disposable devices for filtering fibrin from the serum portion of centrifuged blood samples contained in standard 16mm I.D. collection tubes.
- Standard model is similar to Fisherbrand™ IB™ model, but without the one-way valve
- Use only with glass collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allows direct sampling into automatic laboratory equipment
- Eliminates fibrin, particulate matter and equipment downtime
Specifications
Specifications
| Length (Metric) | 68mm |
| For Use With (Equipment) | Glass collection tubes |
| Diameter (English) Outside | 0.6 in. |
| Length (English) | 2.75 in. |
| Diameter (Metric) Outside | 16mm |
| For Use With (Application) | Used when filtered Serum can be immediately poured off for analysis |
| Product Type | Serum Filter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.