Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Standard Serum Filters

Catalog No. 0268158 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
2 in.
2.75 in.
4 in.
6 in.
Length (Metric):
20mm
68mm
100mm
152mm
Diameter (English) Outside:
0.4 in.
0.5 in.
0.6 in.
Diameter (Metric) Outside:
10.25mm
13mm
16mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Diameter (English) Outside Diameter (Metric) Outside
02-681-58 2 in. 20mm 0.4 in. 10.25mm
02-681-53 4 in. 100mm 0.4 in. 10.25mm
02-681-50 4 in. 100mm 0.5 in. 13mm
02-681-52 6 in. 152mm 0.6 in. 16mm
02-681-51 4 in. 100mm 0.6 in. 16mm
02-681-54 2.75 in. 68mm 0.6 in. 16mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02-681-58

Used when the filtered serum can be immediately poured off for analysis.

Disposable devices for filtering fibrin from the serum portion of centrifuged blood samples contained in standard 10.25mm I.D. collection tubes.

  • Standard model is similar to Fisherbrand™ IB™ model, but without the one-way valve
  • Use only with glass collection tubes
  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allows direct sampling into automatic laboratory equipment
  • Eliminates fibrin, particulate matter and equipment downtime

Specifications

For Use With (Equipment) Glass collection tubes
Length (English) 2 in.
For Use With (Application) Used when filtered Serum can be immediately poured off for analysis
Length (Metric) 20mm
Product Type Serum Filter
Diameter (English) Outside 0.4 in.
Diameter (Metric) Outside 10.25mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.