Learn More
Thermo Scientific™ Standard Single-Use PETG 2-Port Bottle Assembly System
Thermo Scientific™ PETG Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle, a 38 to 430 mm ported closure with one vent filter line and one 1/8-in plugged connection line
$1158.75 - $1816.85
Specifications
Description Polycarbonate standard single-use 2-Port bottle assembly system
Item Description Port 1: PP tube plug port, 12 in (30.48 cm) tubing length, 1/8 x 1/4 (3.2 x 6.2) ID x OD; Port 2: Vent filter, 3 in (7.62 cm) tubing length, 1/8 x 3/8 (3.2 x 9.6) ID x OD; Port 3: PP tube plug, 12 in (30.48 cm) tubing length, 1/8 x 3/8 (3.2 x 9.6) ID x OD
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
SB00008I View Documents Thermo Scientific™
SB00008I
500 mL Pack of 15 for $1,484.06
Add to cart
SB00005I View Documents Thermo Scientific™
SB00005I
125 mL Pack of 10 for $1,158.75
Add to cart
SB00006I View Documents Thermo Scientific™
SB00006I
125 mL Pack of 25 for $1,580.64
Add to cart
SB00007I View Documents Thermo Scientific™
SB00007I
250 mL Pack of 20 for $1,816.85
Add to cart
Description
Description
Key benefits:
  • Designed for a broad range of applications, including bioproduction, life science, and general lab use
  • Helps eliminate setup and post-use cleaning steps required for reuseable containers
  • Helps reduce cross-contamination risks
  • Allows process-specific flexibility with multiple bottle sizes
Assembly details:
  • Bottles are available in polycarbonate (PC) and polyethylene terephthalate glycol (PETG) contact surfaces
  • Each bottle is topped with an assembly including a cap, puck, and tubing
  • Most assemblies contain two ports with C-Flex™ tubing; the 2 L size includes a third port, also with C-Flex tubing
  • One line on each assembly is dedicated to venting
  • The second and third lines are for liquid transfer and include 1/8 in. plug fittings, to be used with your preferred welding or sealing equipment
  • Fluid path of the finished product has been validated according to AAMI TIR33 with a 10-6 sterility assurance level (SAL)
Specifications
Specifications
Polycarbonate standard single-use 2-Port bottle assembly system
Port 1: PP tube plug port, 12 in (30.48 cm) tubing length, 1/8 x 1/4 (3.2 x 6.2) ID x OD; Port 2: Vent filter, 3 in (7.62 cm) tubing length, 1/8 x 3/8 (3.2 x 9.6) ID x OD; Port 3: PP tube plug, 12 in (30.48 cm) tubing length, 1/8 x 3/8 (3.2 x 9.6) ID x OD