Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Water (Fresh Water Substitute), Ricca Chemical

$121.80

Specifications

Chemical Name or Material Standard Water (Fresh Water Substitute)
CAS 10035-04-8, 7791-18-6, 144-55-8
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R348900010F
View Documents
Ricca Chemical Company
R348900010F
10L
Each for $121.80
Add to cart
 
Specifications

Specifications

Standard Water (Fresh Water Substitute)
10035-04-8, 7791-18-6, 144-55-8
Liquid
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit