Learn More
Fisherbrand™ Standing Height Wood Corner/Sink Cabinet
Fisherbrand™ Standing Height Wood Cabinets bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB2414M1
Add to cart
Description
Description
  • Standing Height (35 in. tall)
  • Made from hand selected premium rotary cut maple or rift cut oak veneers.
  • Featuring solid mortise/tenon top frame, doweled, glued and screwed construction.
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Cabinets have a full flush overlay design with squared edges.
  • They are vertically grain matched and are stained inside and out.
Compliance
SEFA 8
Specifications
Specifications
Standing Height
Natural Stain
81.3 cm
88.9 cm
1
2 years
86.9 cm
SEFA 8
32 in.
35 in.
Wood (Maple Veneer)
Corner/Sink Cabinet
34.25 in