Learn More
Fisherbrand™ Standing Height Wood Knee Space Panels for use with Drawer Apron Frames, 30 in. Wide
Fisherbrand™ Wood Knee Space Panels bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB90293XO6
Add to cart
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Knee space panels close off the casework to the call to create a clear sitting or standing space under the work surface
Compliance
SEFA 8
Specifications
Specifications
Chili Stain
2.5 cm
29 in.
Wood (Oak Veneer)
76.2 cm
1 in.
Drawer Apron Frames
73.7 cm
30 in